Floor cleaners, SURE®
Supplier: Diversey
Certificates
About this item
Concentrated, low foaming daily cleaner for all water resistant hard floors. Recommended for all cleaning methods, including bucket and scrubber drier applications.
- Concentrated formulation gives excellent economy of use
- No residues on the floor and no build-up
- Low foaming effective cleaning
- No fragrance or dye
- Contains plant-based ingredients from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications, no safety warnings for user
SURE® floor cleaner is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B.
EU-Ecolabel (EU: NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
EU-Ecolabel (EU: NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
Specifications
- Catalog No:
- 141-1571
- 141-1572
- Environmentally Preferable:
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging