Order Entry
Netherlands
ContactUsLinkComponent
Washroom cleaner and descaler, SURE®
Washroom cleaner and descaler, SURE®
Catalog # 115-3070
Supplier:  Diversey
   Sustainable Materials
   Safer Human and Environmental Health
   Sustainable Packaging
undefined
Washroom cleaner and descaler, SURE®
Catalog # 115-3070
Supplier:  Diversey
   Sustainable Materials
   Safer Human and Environmental Health
   Sustainable Packaging
Specifications
  • Description:
    Washroom cleaner and descaler, 6×1 L
  • Seq:
    01
  • Environmentally Preferable:
    Sustainable Materials,
    Safer Human and Environmental Health,
    Sustainable Packaging
  • Pk:
    6 Bottle
Specifications
About this item
Concentrated, highly effective product for daily and periodical cleaning of water resistant hard surfaces in washrooms.
  • Concentrated formulation gives excellent economy of use
  • Effective cleaning and descaling even in hard water areas
  • Suitable for use on intact chrome and stainless steel surfaces
  • Cleans without leaving residue
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
SURE® washroom cleaner and descaler is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.