Magnetic stirrer heads, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE compound, glass shaft. These gastight stirrer heads are ideal for PTFE-jacketed stirrer shafts from BOLA and are also suitable for shortened stirrer shafts. Suitable for high speeds of up to 1500 min⁻¹.
- Very good chemical resistance, temperature resistance from –200 to +250 °C
- Joint-Cone with nut for easy locking and unlocking of the ground joint
- Adjustable stirrer shaft hight ±40 mm
- Compression fittings for safe fixing of stirrer shaft and optimum power transmission
- No leakage or memory effects due to non-porous, welded rotor
Powerful transmission of up to 90 Ncm. The insertion length of shafts is 95 mm and the drive shaft has an outer Ø of 50 mm.
Powerful transmission of up to 90 Ncm. The insertion length of shafts is 95 mm and the drive shaft has an outer Ø of 50 mm.
Frequently Bought Together