ICP-OES torches for PerkinElmer
Supplier: Agilent
Certificates
About this item
These torches for PerkinElmer ICP-OES are available in a wide range of styles from 0 to 3 slots, depending on the instrument and your application needs.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Torches without slots are recommended for organic solvents. One-slot torches are the standard torch for aqueous solutions. Three-slot torches are typically used for organic analysis.
Frequently Bought Together