STEEL-BRIGHT
Catalog # VELTSB-02
Supplier: VELTEK ASSOCIATES
Specifications
- Omschrijving:STEEL-BRIGHT®, 237 ml
- VPE:24
Specifications
About this item
Designed to remove chemical residues, spotting, and staining on stainless steel, chrome, brass, aluminum, and copper.
This sterile cleaning agent will not rainbow, accumulate a heavy build-up, or leave a powdery residue. Filtered at 0,2 µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.