Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-CD90Thy-1 Rat Antibody
Anti-CD90Thy-1 Rat Antibody
Anti-CD90Thy-1 Rat Antibody
Catalog # APOSBITP2024-1MG
Supplier:
Apollo Scientific
Expand image
Anti-CD90Thy-1 Rat Antibody
Catalog # APOSBITP2024-1MG
Supplier:
Apollo Scientific
Specifications
Antigen symbol:
CD90Thy-1
Host:
Rat
Reactivity:
Mouse
Pk:
1 mg
Specifications
About this item
Product Details
Product Details