Ga naar Rubriek
Ga naar Hoofdinhoud
Ga naar Voettekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Mouse recombinant IP-10
Mouse recombinant IP-10
Mouse recombinant IP-10
Catalog # APOSBITC2020-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Mouse recombinant IP-10
Catalog # APOSBITC2020-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Mouse
Protein synonyms:
CXCL10
Protein/peptide name:
IP-10
Specifications
About this item
Product Details
Product Details