Human recombinant ENA-78
Catalog # APOSBITC2004-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Epithelial Neutrophil-Activating Protein 78, CXCL5
- Protein/peptide name:ENA-78