Order Entry
Netherlands
ContactUsLinkComponent
1,3-Diethynyl-5-(trifluoromethoxy)benzene
1,3-Diethynyl-5-(trifluoromethoxy)benzene
Catalog # APOSPC50556-5G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Diethynyl-5-(trifluoromethoxy)benzene
Catalog # APOSPC50556-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :210.152 g/mol
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
Formula:C₁₁H₅F₃O