
[(1-Cyclopropyl-1H-pyrazol-4-yl)methyl](methyl)amine dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2514953-01-4
:
Formula:C₈H₁₃N₃.₂ClH
:224.131 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together