Enniatin Complex
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
- CAS-nummer:11113-62-5
- MDL-nummer:MFCD00214209
- Smelten Pt:120…128 °C
- Smelten Pt:120-128 °C
- Temperatuur bij opslag:Freezer
- UN:3462
- ADR:6.1,III
5 Opties Hieronder Beschikbaar
Product Details & Documents
- CAS-nummer:11113-62-5
- MDL-nummer:MFCD00214209
- Smelten Pt:120…128 °C
- Smelten Pt:120-128 °C
- Temperatuur bij opslag:Freezer
- UN:3462
- ADR:6.1,III
Product Family Options
Productinformatie
- Pack typePkBeschikbaarheidPrice
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



