
6-deoxy-6-((2-carboxyethyl)disulphide)-heptakis(6-deoxy-6-(2-carboxyethyl)thio)-gamma-cyclodextrin sodium salt
Catalog # APOSBICY1064-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient