

2-Hydroxy-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50650-59-4
:
Formula:C₆H₄F₃NO
MDL Number:MFCD00153195
:163.1 g/mol
Melting Pt:158…165 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together