
4-(Bromomethyl)-1-fluoro-2-nitrobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15017-52-4
:
Formula:C₇H₅BrFNO₂
MDL Number:MFCD03412241
:234.0225 g/mol
Melting Pt:55…57 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together