Order Entry
Netherlands
ContactUsLinkComponent
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine 97%
Irritant
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine 97%
Catalog # APOSPC9484-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine 97%
Catalog # APOSPC9484-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:140…142 °C
 :373.35 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:250714-63-7
Formula:C₁₆H₁₄F₃NO₄S
MDL Number:MFCD00173005