Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
2,3-Difluoro-5-(trifluoromethyl)pyridine
Catalog # APOSPC9478-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Difluoro-5-(trifluoromethyl)pyridine
Catalog # APOSPC9478-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :183.08 g/mol
Boiling Pt:104 °C
Density:1.47 g/cm³ (25 °C)
Flash Pt:>100 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:89402-42-6
Formula:C₆H₂F₅N
MDL Number:MFCD04972837