
1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338982-15-3
:
Formula:C₈H₅F₃N₂O₂S
MDL Number:MFCD01814674
:250.2 g/mol
Melting Pt:255…258 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together