[1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:318469-22-6
:
Formula:C₁₂H₁₁F₃N₂O₂
MDL Number:MFCD00172652
:272.23 g/mol
Melting Pt:80…82 °C
Storage Temperature:Ambient
Frequently Bought Together