4-Methyl-5-[1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazol-5-yl]-4H-1,2,4-triazole-3-thiol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338747-58-3
:
Formula:C₁₀H₈F₃N₅S₂
MDL Number:MFCD01444047
:319.33 g/mol
Melting Pt:306…309 °C
Storage Temperature:Ambient
Frequently Bought Together