
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine
Catalog # APOSPC9218-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:307.44 g/mol
Melting Pt:37…39 °C
:GHS07
Storage Temperature:Ambient
CAS Number:505084-56-0
Formula:C₆H₂ClF₃IN
MDL Number:MFCD08741352