
4-Fluoro-3-iodobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:403-18-9
:
Formula:C₇H₄FIO₂
MDL Number:MFCD03788556
:266.01 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together