Order Entry
Netherlands
ContactUsLinkComponent
[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98%
[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98%
Catalog # APOSPC906499-1G
Supplier:  Apollo Scientific
CAS Number:  
[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98%
Catalog # APOSPC906499-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :325.05 g/mol
Storage Temperature:Ambient
CAS Number:1046786-08-6
Formula:C₉H₁₁BF₇NOS