
(2-Bromoethyl)diphenylsulphonium trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:247129-85-7
:
Formula:C₁₅H₁₄BrF₃O₃S₂
MDL Number:MFCD12546049
:443.29 g/mol
Melting Pt:85…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together