
Octafluoroadiponitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:376-53-4
:
Formula:C₆F₈N₂
MDL Number:MFCD00155894
:252.07 g/mol
Boiling Pt:61…63 °C
Density:1.4304 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together