N-[4-Chloro-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:167415-21-6
:
Formula:C₉H₃ClF₆N₂O₃
MDL Number:MFCD08689680
:336.58 g/mol
Melting Pt:100…102 °C
Storage Temperature:Ambient
Frequently Bought Together