Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Catalog # APOSPC8571-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Catalog # APOSPC8571-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.67 g/mol
Melting Pt:130…131 °C
Storage Temperature:Ambient
CAS Number:952183-37-8
Formula:C₁₃H₉ClFNO₂
MDL Number:MFCD09817431