Order Entry
Netherlands
ContactUsLinkComponent
N-(2-Bromo-4-fluorophenyl)thiourea
N-(2-Bromo-4-fluorophenyl)thiourea
Catalog # APOSPC8481-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(2-Bromo-4-fluorophenyl)thiourea
Catalog # APOSPC8481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :249.11 g/mol
Melting Pt:163…165 °C
Storage Temperature:Ambient
CAS Number:952182-71-7
Formula:C₇H₆BrFN₂S
MDL Number:MFCD09027194