5-(2,4-Dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321848-48-0
:
Formula:C₁₂H₇Cl₂F₃N₂O₂
MDL Number:MFCD01935965
:339.1 g/mol
Melting Pt:128…129 °C
Storage Temperature:Ambient
Frequently Bought Together