Order Entry
Netherlands
ContactUsLinkComponent
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 97%
Irritant
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 97%
Catalog # APOSPC8416-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 97%
Catalog # APOSPC8416-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :288.66 g/mol
Melting Pt:66…68 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:80783-47-7
Formula:C₁₂H₈ClF₃N₂O
MDL Number:MFCD01315057