
2-Bromo-2'-fluoro-5'-(trifluoromethyl)acetophenone 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:202664-36-6
:
Formula:C₉H₅BrF₄O
MDL Number:MFCD03094506
Melting Pt:15…17 °C
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together