Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-5-[(2-methyl-1H-imidazol-1-yl)sulphonyl]benzotrifluoride
3-Bromo-5-[(2-methyl-1H-imidazol-1-yl)sulphonyl]benzotrifluoride
Catalog # APOSPC8277-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-5-[(2-methyl-1H-imidazol-1-yl)sulphonyl]benzotrifluoride
Catalog # APOSPC8277-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:951884-57-4
Formula:C₁₁H₈BrF₃N₂O₂S
MDL Number:MFCD09801016