
4-(Trifluoromethoxy)benzotrifluoride 98%
Catalog # APOSPC8213-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.11 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:80258-33-9
Formula:C₈H₄F₆O
MDL Number:MFCD00190125