2-[4-Chlorobutyrylamino)phenoxy]-3-chloro-5-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339014-65-2
:
Formula:C₁₆H₁₃Cl₂F₃N₂O₂
MDL Number:MFCD00664603
:393.20 g/mol
Melting Pt:152…154 °C
Storage Temperature:Ambient
Frequently Bought Together