

3-(Trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2338-76-3
:
Formula:C₉H₆F₃N
MDL Number:MFCD00001913
:185.15 g/mol
Boiling Pt:92…93 °C/4mmHg
Density:1.187 g/cm³ (25 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:219-053-8
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together