

2-Bromo-4'-(trifluoromethyl)acetophenone 97%
:383-53-9
:
:C₉H₆BrF₃O
:MFCD00126489
:267.0400 g/mol
:46…56 °C
:3261
:8,III
:GHS05
:GHS07
:Refrigerator
Frequently Bought Together
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
1-Pentanol ≥98%
: Thermo Fisher Scientific
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Butyl acetate ACS
: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
: MACRON AVANTOR BRAND