Order Entry
Netherlands
ContactUsLinkComponent
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate 97%
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate 97%
Catalog # APOSPC7612-5G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate 97%
Catalog # APOSPC7612-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :356.41 g/mol
Storage Temperature:Ambient
CAS Number:59643-43-5
Formula:C₁₅H₂₃F₃O₄S
MDL Number:MFCD00229854