Order Entry
Netherlands
ContactUsLinkComponent
cis-1-Iodo-2-(pentafluoroethyl)cyclohexane 97%
cis-1-Iodo-2-(pentafluoroethyl)cyclohexane 97%
Catalog # APOSPC7563-100MG
Supplier:  Apollo Scientific
CAS Number:  
cis-1-Iodo-2-(pentafluoroethyl)cyclohexane 97%
Catalog # APOSPC7563-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :328.07 g/mol
Boiling Pt:70 °C/6mmHg
Density:1.78 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:38787-68-7
Formula:C₈H₁₀F₅I
MDL Number:MFCD01862037