

2-(Trifluoromethoxy)phenyl isocyanate 92%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:182500-26-1
:
Formula:C₈H₄F₃NO₂
MDL Number:MFCD00134160
:203.12 g/mol
Boiling Pt:52…55 °C/15mmHg
Density:1.35 g/cm³ (25 °C)
Flash Pt:60 °C
Storage Temperature:Refrigerator
UN:3080
ADR:6.1+3,II
:GHS02
:GHS06
Frequently Bought Together