
2-Bromo-3'-(trifluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:237386-01-5
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD03094425
:283.0400 g/mol
Boiling Pt:86 °C
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together