
1-Amino-2,2,2-trifluoroethane hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:373-88-6
:
Formula:C₂H₅ClF₃N
MDL Number:MFCD00012875
:135.52 g/mol
Melting Pt:220…222 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together