
7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86393-33-1
:
Formula:C₁₃H₉ClFNO₃
MDL Number:MFCD00792458
:281.6669 g/mol
Melting Pt:242…245 °C
EINECS:405-050-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together