
2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine
Catalog # APOSPC6739-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:199.54 g/mol
Boiling Pt:35…37 °C/25mmHg
Density:1.5 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
CAS Number:628692-22-8
Formula:C₆H₂ClF₄N
MDL Number:MFCD04038326