(2-Amino-5-chlorophenyl)(2,6-difluorophenyl)methanone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:28910-83-0
:
Formula:C₁₃H₈ClF₂NO
MDL Number:MFCD06659048
:267.6600 g/mol
Melting Pt:103…105 °C
Storage Temperature:Ambient
Frequently Bought Together