
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:282523-49-3
:
Formula:C₁₂H₈ClF₃N₂O₃
MDL Number:MFCD00243689
:320.66 g/mol
Melting Pt:177…178 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together