Order Entry
Netherlands
ContactUsLinkComponent
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoicacid, methyl ester 95%
Corrosive
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoicacid, methyl ester 95%
Catalog # APOSPC6220J-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoicacid, methyl ester 95%
Catalog # APOSPC6220J-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :676.13 g/mol
Boiling Pt:193…196 °C
Density:1.6 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1760
ADR:8,III
 :GHS05
CAS Number:39187-47-8
Formula:C₁₃H₃F₂₃O₅
MDL Number:MFCD00054724