Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol 99%
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol 99%
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol 99%
APOSPC6085-100G
:
Apollo Scientific
:
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol 99%
APOSPC6085-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
264.09 g/mol
:
140…143 °C
:
1.59 g/cm³ (25 °C)
:
73 °C
:
218-050-9
:
GHS07
:
Ambient
:
2043-47-2
:
C₆H₅F₉O
:
MFCD00039543