
2-Chloro-6-(trifluoromethyl)nicotinamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:386704-05-8
:
Formula:C₇H₄ClF₃N₂O
MDL Number:MFCD01862659
:224.57 g/mol
Melting Pt:198…200 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together