
1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carbonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175137-36-7
:
Formula:C₁₁H₅ClF₃N₃O₃
MDL Number:MFCD00173866
:319.63 g/mol
Melting Pt:77…79 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together