
2'-(4-Acetophenoxy)-3'-chloro-5'-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:217186-15-7
:
Formula:C₁₄H₉CIF₃NO₂
MDL Number:MFCD00170137
:GHS07
Storage Temperature:Ambient
Frequently Bought Together