Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-4-nitro-5-(trifluoromethyl)aniline ≥98%
2-Bromo-4-nitro-5-(trifluoromethyl)aniline ≥98%
Catalog # APOSPC540082-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-4-nitro-5-(trifluoromethyl)aniline ≥98%
Catalog # APOSPC540082-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :285.0181 g/mol
Storage Temperature:Ambient
CAS Number:1190198-31-2
Formula:C₇H₄BrF₃N₂O₂
MDL Number:MFCD12922693